missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kelch Domain Containing 7A Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98170DL594
This item is not returnable.
View return policy
Description
Kelch Domain Containing 7A Polyclonal antibody specifically detects Kelch Domain Containing 7A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Kelch Domain Containing 7A | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 127707 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| Kelch Domain-Containing Protein 7A, KLHDC7A, RP11-422P22.2 | |
| Produced in rabbits immunized with E. coli-derived Human Kelch Domain Containing 7A fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction