Learn More
Hamilton™ Custom Kel-F Hub Needles
Product Code. 11580072
Product Code. 11580072 Supplier Hamilton™ Supplier No. 775004
Only null left
Add to basket
Add to basket
Design controlled by a series of universal standards which guarantees compatibility between manufacturers.
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style
  • Autoclavable
  • Gas sterilizable
  • For Kel-F hubs
  • Custom length
  • Stainless steel Luer tip