Learn More
Duran™ Keck Hose Clamps
Control the amount of a substance flowing through the hose using only one finger
Brand: Duran™ 286338101
This item is not returnable.
View return policy
Description
- Amount of a substance flowing through the hose can be controlled precisely using only one finger
- Thermoplastic polyester (PBT) clamps are temperature resistant up to 200°C
- Autoclavable