Learn More
Duran™ Keck Hose Clamps
Control the amount of a substance flowing through the hose using only one finger
Brand:  Duran™ 286338204
Product Code. 10101691
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Amount of a substance flowing through the hose can be controlled precisely using only one finger
  • Thermoplastic polyester (PBT) clamps are temperature resistant up to 200°C
  • Autoclavable