Learn More
Duran™ Keck Conical Ground Clamps
Made of temperature resistant polyacetal (POM)
Brand:  Duran™ 286336803
Product Code. 10507281
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • KECK™ Clips for conical joints are available in sizes NS 10 up to NS 45
  • Made of temperature resistant polyacetal (POM) Good spring properties
  • Colour coded by size
  • No risk of scratching the glass
  • Will not corrode or bend out of shape