Learn More
Duran™ Keck Conical Ground Clamps
Made of temperature resistant polyacetal (POM)
Brand: Duran™ 286336203
This item is not returnable.
View return policy
Description
- KECK™ Clips for conical joints are available in sizes NS 10 up to NS 45
- Made of temperature resistant polyacetal (POM) Good spring properties
- Colour coded by size
- No risk of scratching the glass
- Will not corrode or bend out of shape