Learn More
DWK Life Sciences KECK™ Clip Assortment, for conical joints, from stainless steel (1.4310, blank)
The KECK™ clamp is used to secure a standard ground joint connection.
Brand:  DWK Life Sciences 290300408
Code : NEW
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10719474
  • £45.00 / Pack of 10
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
Specifications
Specifications
10
Secure Glass-to-Glass Joints
NS 12
Stainless Steel (1.4310, Blank)
Secure glass-to-glass joints