Learn More
Invitrogen™ KEAP1 Monoclonal Antibody (7G4B10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517107
Description
MA5-17107 targets KEAP1 in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17107 immunogen is purified recombinant fragment of human KEAP1 (amino acids: 380-624 ) expressed in E. Coli. MA5-17107 detects KEAP1 which has a predicted molecular weight of approximately 69.7kDa.
This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.
Specifications
| KEAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q14145 | |
| Keap1 | |
| Purified recombinant fragment of human KEAP1 (amino acids: 380-624 ) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 7G4B10 | |
| Unconjugated | |
| Keap1 | |
| cytosolic inhibitor of Nrf2; INrf2; Keap1; kelch like ECH associated protein 1; Kelch-like ECH-associated protein 1; kelch-like family member 19; kelch-like protein 19; Kiaa0132; KLHL19; mKIAA0132; NRF2 cytosolic inhibitor; ring canal protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 9817 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.