missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KEAP1 Monoclonal Antibody (297CT6.1.6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537537
This item is not returnable.
View return policy
Description
KEAP1 Monoclonal Antibody for Western Blot
This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.
Specifications
| KEAP1 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q14145 | |
| Keap1 | |
| KLH conjugated synthetic peptide between 422-449 amino acids from human KEAP1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| Western Blot | |
| 297CT6.1.6 | |
| Unconjugated | |
| Keap1 | |
| cytosolic inhibitor of Nrf2; INrf2; Keap1; kelch like ECH associated protein 1; Kelch-like ECH-associated protein 1; kelch-like family member 19; kelch-like protein 19; Kiaa0132; KLHL19; mKIAA0132; NRF2 cytosolic inhibitor; ring canal protein | |
| Mouse | |
| Euglobulin Precipitation | |
| RUO | |
| 9817 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction