missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keap1 Antibody (OTI1B4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-03319
This item is not returnable.
View return policy
Description
Keap1 Monoclonal specifically detects Keap1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Keap1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
| KEAP1 | |
| Full length human recombinnt protein of human KEAP1(NP_987096) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI1B4 | |
| Unconjugated | |
| INRF2, INrf2MGC9454, kelch-like ECH-associated protein 1, Kelch-like protein 19, KIAA0132MGC4407, KLHL19Cytosolic inhibitor of Nrf2, MGC10630, MGC1114, MGC20887 | |
| Mouse | |
| 69.5 kDa | |
| 0.1 mL | |
| Stem Cell Markers | |
| 9817 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction