missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keap1 Antibody (7G4B10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00 - £420.00
Specifications
| Antigen | Keap1 |
|---|---|
| Clone | 7G4B10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18700974
|
Novus Biologicals
NBP2-37431 |
0.1 mL |
£420.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674246
|
Novus Biologicals
NBP2-37431-0.025ml |
0.025 mL |
£196.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Keap1 Monoclonal specifically detects Keap1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Keap1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| INRF2, INrf2MGC9454, kelch-like ECH-associated protein 1, Kelch-like protein 19, KIAA0132MGC4407, KLHL19Cytosolic inhibitor of Nrf2, MGC10630, MGC1114, MGC20887 | |
| KEAP1 | |
| IgG1 | |
| Affinity Purified |
| 7G4B10 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Mouse | |
| Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 9817 | |
| Purified recombinant fragment of human KEAP1 (AA: 380-624 ) expressed in E. Coli. | |
| Primary | |
| 69.7 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title