missing translation for 'onlineSavingsMsg'
Learn More

Keap1 Antibody (333116) [Biotin], Novus Biologicals™

Product Code. 30735377 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735377 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735377 Supplier Novus Biologicals Supplier No. FAB3024B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Keap1 Monoclonal antibody specifically detects Keap1 in Human,Mouse,Rat samples. It is validated for Western Blot,Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen Keap1
Applications Western Blot, Gene Knock-Out
Classification Monoclonal
Clone 333116
Conjugate Biotin
Formulation PBS
Gene Alias INRF2, INrf2MGC9454, kelch-like ECH-associated protein 1, Kelch-like protein 19, KIAA0132MGC4407, KLHL19Cytosolic inhibitor of Nrf2, MGC10630, MGC1114, MGC20887
Host Species Mouse
Immunogen E. coli-derived recombinant human Keap1, Ala90-Ile250, Accession # Q14145
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9817
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.