missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ kdsB Polyclonal Antibody
GREENER_CHOICE

Product Code. 17919062
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17919062 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17919062 Supplier Invitrogen™ Supplier No. PA5144388

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

kdsB Polyclonal Antibody for Western Blot, ELISA

Activates KDO (a required 8-carbon sugar) for incorporation into bacterial lipopolysaccharide in Gram-negative bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen kdsB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene kdsB
Gene Accession No. B5YT52
Gene Alias 3-deoxy-manno-octulosonate cytidylyltransferase kdsB ECH74115_1079
Gene Symbols kdsB
Host Species Rabbit
Immunogen Recombinant Escherichia coli O157:H7 (strain EC4115/EHEC) kdsB protein (1-248aa ).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75170992
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.