missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KDM6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551685
This item is not returnable.
View return policy
Description
Immunogen sequence: IGNNHITGSG SNGNVPYLQR NALTLPHNRT NLTSSAEEPW KNQLSNSTQG LHKGQSSHSA GPNGERPLSS TGPSQHLQAA GSGIQNQNGH PTLPSNSVTQ GAALNHLSSH TATSGGQQGI TLTKESKPSG NILTVPETSR HT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
This gene is located on the X chromosome and is the corresponding locus to a Y-linked gene which encodes a tetratricopeptide repeat (TPR) protein. The encoded protein of this gene contains a JmjC-domain and catalyzes the demethylation of tri/dimethylated histone H3.
Specifications
| KDM6A | |
| Polyclonal | |
| Unconjugated | |
| KDM6A | |
| 4lysine (K)-specific demethylase 6A; bA386N14.2; bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeat protein (UTX)); histone demethylase UTX; KABUK2; Kdm6a; lysine (K)-specific demethylase 6A; lysine demethylase 6A; lysine-specific demethylase 6A; lysine-specific demethylase 6A-like protein; NEWGENE_1565481; ubiqiutous TPR motif protein; ubiquitous TPR motif protein UTX; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat protein X-linked; ubiquitously transcribed tetratricopeptide repeat, X chromosome; ubiquitously transcribed TPR protein on the X chromosome; Ubiquitously transcribed X chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR gene on the X chromosome; Ubiquitously-transcribed TPR protein on the X chromosome; Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein; UTX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7403 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15550 | |
| KDM6A | |
| Recombinant protein corresponding to Human KDM6A. Recombinant protein control fragment (Product #RP-101025). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction