missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KDM6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115463
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KDM6A / UTX.
This gene is located on the X chromosome and is the corresponding locus to a Y-linked gene which encodes a tetratricopeptide repeat (TPR) protein. The encoded protein of this gene contains a JmjC-domain and catalyzes the demethylation of tri/dimethylated histone H3.
Specifications
| KDM6A | |
| Polyclonal | |
| Unconjugated | |
| KDM6A | |
| 4lysine (K)-specific demethylase 6A; bA386N14.2; bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeat protein (UTX)); histone demethylase UTX; KABUK2; Kdm6a; lysine (K)-specific demethylase 6A; lysine demethylase 6A; lysine-specific demethylase 6A; lysine-specific demethylase 6A-like protein; NEWGENE_1565481; ubiqiutous TPR motif protein; ubiquitous TPR motif protein UTX; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat protein X-linked; ubiquitously transcribed tetratricopeptide repeat, X chromosome; ubiquitously transcribed TPR protein on the X chromosome; Ubiquitously transcribed X chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR gene on the X chromosome; Ubiquitously-transcribed TPR protein on the X chromosome; Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein; UTX | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 103689969, 22289, 7403 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O15550, O70546 | |
| KDM6A | |
| A synthesized peptide derived from human KDM6A(Accession O15550), corresponding to amino acid residues H1029-F1079. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction