missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM5B Mouse anti-Human, Clone:7H3D7, Abnova™
Mouse monoclonal antibody raised against recombinant human KDM5B.
Brand: Abnova MAB16745.100ug
This item is not returnable.
View return policy
Specifications
| KDM5B | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| KDM5B | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 7H3D7 | |
| jumonji, AT rich interactive domain 1B | |
| In PBS (0.05% sodium azide) | |
| FLJ10538, FLJ12459, FLJ12491, FLJ16281, FLJ23670, KDM5B, PLU-1, PUT1, RBBP2H1A | |
| Recombinant protein corresponding to amino acid 231-319 of human KDM5B from E. coli. | |
| 10765 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction