missing translation for 'onlineSavingsMsg'
Learn More

KDM4D-IN-1, MedChemExpress

Product Code. 16474726
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474726 100 mg 100mg
16450037 50 mg 50mg
16436377 5 mg 5mg
16420949 1 mg 1mg
16413648 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16474726 Supplier Medchem Express Supplier No. HY101928
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress KDM4D-IN-1 is a new histone lysine demethylase 4D (KDM4D) inhibitor with an IC50 value of 0.41±0.03 μM.

KDM4D-IN-1 is a new histone lysine demethylase 4D (KDM4D) inhibitor with an IC50 value of 0.41±0.03 μM.

Specifications

Chemical Name or Material KDM4D-IN-1
CAS 2098902-68-0
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C11H7N5O
Quantity 100 mg
Solubility Information DMSO : 7.14 mg/mL (31.70 mM; Need ultrasonic)
SMILES OC1=NC2=C(C#N)C(C)=NN2C3=C1C=CC=N3
Molecular Weight (g/mol) 225.21
Formula Weight 225.21
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.