missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM3A/JMJD1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12835-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognize the N-term of KDM3A (JMJD1A).
This gene encodes a zinc finger protein that contains a jumonji domain and may play a role in hormone-dependent transcriptional activation. Alternative splicing results in multiple transcript variants.Specifications
| KDM3A/JMJD1A | |
| Polyclonal | |
| Unconjugated | |
| KDM3A | |
| JHDM2A, JHMD2A, JMJD1, JMJD1A, KDM3A, KIAA0742, lysine (K) demethylase 3A, Lysine specific demethylase 3A, TSGA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 104263, 55818 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6PCM1, Q9Y4C1 | |
| KDM3A | |
| KDM3A,JMJD1A Fusion Protein Ag3581 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction