missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM2B Antibody (3B7D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52466-0.025mg
This item is not returnable.
View return policy
Description
KDM2B Monoclonal specifically detects KDM2B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| KDM2B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| [Histone-H3]-lysine-36 demethylase 1B, CXXC2F-box protein FBL10, CXXC-type zinc finger protein 2, Fbl10, F-box and leucine-rich repeat protein 10JEMMA (Jumonji domain, EMSY-interactor, methyltransferase motif) protein, FBXL10, JHDM1BF-box/LRR-repeat protein 10, JmjC domain-containing histone demethylation protein 1B, jumonji C domain-containing histone demethylase 1B, Jumonji domain-containing EMSY-interactor methyltransferase motif protein, lysine (K)-specific demethylase 2B, lysine-specific demethylase 2B, PCCX2EC 1.14.11.27, protein containing CXXC domain 2, Protein JEMMA, Protein-containing CXXC domain 2 | |
| Mouse | |
| 152.6 kDa | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 3B7D11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| KDM2B | |
| Purified recombinant fragment of human FBXL10 (AA: 457-555) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| 84678 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction