missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KDM2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540421
This item is not returnable.
View return policy
Description
Peptide sequence: RIRYSQRLRG TMRRRYEDDG ISDDEIEGKR TFDLEEKLHT NKYNANFVTF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
FBXL11 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box). The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. FBXL11 belongs to the Fbls class and, in addition to an F-box, contains at least 6 highly degenerated leucine-rich repeats.
Specifications
| KDM2A | |
| Polyclonal | |
| Unconjugated | |
| KDM2A | |
| [Histone-H3]-lysine-36 demethylase 1A; 100043628; 5530401A10Rik; AA589516; AW536790; CXXC8; CXXC-type zinc finger protein 8; DKFZp434M1735; FBL11; FBL7; F-box and leucine-rich repeat protein 11; F-box protein containing leucine repeats; F-box protein FBL7; F-box protein Lilina; F-box/LRR-repeat protein 11; Fbxl11; FLJ00115; FLJ46431; Gm4560; Jhdm1; JHDM1A; JmjC domain-containing histone demethylation protein 1A; jumonji C domain-containing histone demethylase 1A; Kdm2a; KIAA1004; lalina; LILINA; lysine (K)-specific demethylase 2A; lysine demethylase 2A; lysine-specific demethylase 2A; SF-KDM2A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22992 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y2K7 | |
| KDM2A | |
| Synthetic peptide directed towards the N-terminal of human FBXL11 (aa 7-56). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction