missing translation for 'onlineSavingsMsg'
Learn More

KDM2A/FBXL11 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18428687 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428687 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428687 Supplier Novus Biologicals Supplier No. NBP178305JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KDM2A/FBXL11 Polyclonal antibody specifically detects KDM2A/FBXL11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen KDM2A/FBXL11
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias [Histone-H3]-lysine-36 demethylase 1A, CXXC8F-box protein FBL7, CXXC-type zinc finger protein 8, FBL11DKFZp434M1735, FBL7F-box protein Lilina, F-box and leucine-rich repeat protein 11DKFZP434M1735, F-box protein FBL11, FBXL11FLJ46431, FLJ00115, JHDM1AF-box/LRR-repeat protein 11, JmjC domain-containing histone demethylation protein 1A, jumonji C domain-containing histone demethylase 1A, KIAA1004EC 1.14.11.27, LILINA, lysine (K)-specific demethylase 2A, lysine-specific demethylase 2A, SF-KDM2A
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human KDM2A/FBXL11 protein (between residues 350-500) [UniProt Q9Y2K7]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 22992
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.