missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM2A/FBXL11 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78305AF405
This item is not returnable.
View return policy
Description
KDM2A/FBXL11 Polyclonal specifically detects KDM2A/FBXL11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| KDM2A/FBXL11 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Q9Y2K7 | |
| KDM2A | |
| A synthetic peptide made to an N-terminal portion of the human KDM2A/FBXL11 protein (between residues 350-500) [UniProt Q9Y2K7] | |
| 0.1 mL | |
| Cancer, Epigenetics | |
| 22992 | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| [Histone-H3]-lysine-36 demethylase 1A, CXXC8F-box protein FBL7, CXXC-type zinc finger protein 8, FBL11DKFZp434M1735, FBL7F-box protein Lilina, F-box and leucine-rich repeat protein 11DKFZP434M1735, F-box protein FBL11, FBXL11FLJ46431, FLJ00115, JHDM1AF-box/LRR-repeat protein 11, JmjC domain-containing histone demethylation protein 1A, jumonji C domain-containing histone demethylase 1A, KIAA1004EC 1.14.11.27, LILINA, lysine (K)-specific demethylase 2A, lysine-specific demethylase 2A, SF-KDM2A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction