missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KDM1B Recombinant Rabbit Monoclonal Antibody (23GB6265)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557451
This item is not returnable.
View return policy
Description
KDM1B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The KDM1B gene, also known as lysine demethylase 1B, or LSD2/AOF1, encodes a histone demethylase that specifically targets H3K4me1/2 marks, which are critical for regulating gene expression. KDM1B is an epigenetic modifier that plays a pivotal role in modulating chromatin structure and, consequently, gene transcription. Unlike its homolog KDM1A, KDM1B associates mainly with the gene bodies of actively transcribed genes rather than promoters, indicating its unique role in transcriptional regulation. KDM1B is implicated in various physiological processes and pathophysiological conditions, including tumor progression where it enhances somatic cell reprogramming and promotes the enrichment of tumor stem cells. Its demethylation activity against histones H3K4 and H3K9 underscores its influence on gene silencing and activation pathways, marking it as a significant target in cancer research and potential therapeutic interventions. Understanding the structural and functional nuances of KDM1B contributes to broader insights into epigenetic regulation mechanisms and their implications in disease states.
Specifications
| KDM1B | |
| Recombinant Monoclonal | |
| Unconjugated | |
| KDM1B | |
| 4632428N09Rik; AI482520; amine oxidase (flavin containing) domain 1; amine oxidase, flavin containing 1; Aof1; bA204B7.3; C6orf193; dJ298J15.2; Flavin-containing amine oxidase domain-containing protein 1; KDM1B; LSD2; lysine (K)-specific demethylase 1B; lysine demethylase 1B; lysine-specific histone demethylase 1B; Lysine-specific histone demethylase 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 221656, 306819 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6265 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NB78 | |
| KDM1B | |
| A synthesized peptide derived from human LSD2 / AOF1 (aa 75-136). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction