missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM1B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67886-1-IG-20UL
This item is not returnable.
View return policy
Description
The KDM1B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KDM1B. This antibody recognizes human, mouse, rat antigen. The KDM1B antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.Specifications
| KDM1B | |
| Monoclonal | |
| 1000 μg/mL | |
| C6orf193; DKFZp686I0412; FLJ33898; FLJ34109; FLJ43328; bA204B7.3; dJ298J15.2 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 92 kDa | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), ELISA | |
| 1E1A3 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 221656 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction