missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20813-1-AP-20UL
This item is not returnable.
View return policy
Description
LSD1 is a histone demethylase that specifically demethylates ′Lys-4′ of histone H3, a specific tag for epigenetic transcriptional activation, thereby acting as a corepressor. LSD1 contains a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. It acts by oxidizing the substrate by FAD to generate the corresponding imine that is subsequently hydrolyzed. LSD1 demethylates both mono- and tri-methylted ′Lys-4′ of histone H3. This protein may play a role in the repression of neuronal genes. Alone, it is unable to demethylate H3 ′Lys-4′ on nucleosomes and requires the presence of RCOR1/CoREST to achieve such activity. It may also demethylate ′Lys-9′ of histone H3, a specific tag for epigenetic transcriptional repression, thereby leading to derepression of androgen receptor target genes. Mutations affecting this gene can result in cleft palate, psychomotor retardation, and distintive facial features.Specifications
| KDM1 | |
| Polyclonal | |
| Unconjugated | |
| KDM1A | |
| AOF2, BHC110, KDM1, KDM1A, KIAA0601, LSD1, lysine (K) demethylase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23028, 99982 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60341, Q6ZQ88 | |
| Kdm1a | |
| KDM1 Fusion Protein Ag14778 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction