missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KDELR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599316
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KDELR3.
This gene encodes a member of the KDEL endoplasmic reticulum protein retention receptor family. Retention of resident soluble proteins in the lumen of the endoplasmic reticulum (ER) is achieved in both yeast and animal cells by their continual retrieval from the cis-Golgi, or a pre-Golgi compartment. Sorting of these proteins is dependent on a C-terminal tetrapeptide signal, usually lys-asp-glu-leu (KDEL) in animal cells, and his-asp-glu-leu (HDEL) in S. cerevisiae. This process is mediated by a receptor that recognizes, and binds the tetrapeptide-containing protein, and returns it to the ER. In yeast, the sorting receptor encoded by a single gene, ERD2, is a seven-transmembrane protein. Unlike yeast, several human homologs of the ERD2 gene, constituting the KDEL receptor gene family, have been described. KDELR3 was the third member of the family to be identified. Alternate splicing results in multiple transcript variants.
Specifications
| KDELR3 | |
| Polyclonal | |
| Unconjugated | |
| KDELR3 | |
| AI173274; ER lumen protein retaining receptor 3; ER lumen protein-retaining receptor 3; ERD23; ERD2L3; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 3; KDEL endoplasmic reticulum protein retention receptor 3; KDEL receptor 3; Kdelr3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 105785, 11015 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43731, Q8R1L4 | |
| KDELR3 | |
| A synthesized peptide derived from human KDELR3(Accession O43731), corresponding to amino acid residues V62-S112. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction