missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDELR2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-85141-0.1ML
This item is not returnable.
View return policy
Description
KDELR2 Polyclonal specifically detects KDELR2 in Human samples. It is validated for Western Blot.
Specifications
| KDELR2 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 11014 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| ELP-1(Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2, ER lumen protein retaining receptor 2, ERD2.2FLJ45532, ERD-2-like protein, ERD2-like protein 1, KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2, KDEL endoplasmic reticulum protein retention receptor 2, KDEL receptor 2 | |
| The immunogen is a synthetic peptide directed towards the middle region of Human KDELR2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction