Learn More
Invitrogen™ KDELR1 Monoclonal Antibody (KR-10), APC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545149
Product Code. 17856722
  • £368.00 / 100µg
Description
1 μg/mL was sufficient for detection of KDEL receptor in 20 μg monkey Vero cell lysate by colorimetric immunoblot analysis using Goat Anti-Mouse IgG:AP as the secondary. Detects approximately 25kDa.
KDELR1 (KDEL Endoplasmic Reticulum Protein Retention Receptor 1) is a Protein Coding gene. Diseases associated with KDELR1 include Syndromic X-Linked Intellectual Disability 94 and Myopathy, Centronuclear, 1. Among its related pathways are Transport to the Golgi and subsequent modification and Activation of cAMP-Dependent PKA. Gene Ontology (GO) annotations related to this gene include ER retention sequence binding and KDEL sequence binding. An important paralog of this gene is KDELR2.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KDELR1
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.2
P24390, P33946, Q569A6, Q99JH8
Mouse
Protein G
RUO
100349892, 100520962, 100750912, 101122047, 10945, 361577, 484406, 618184, 68137
4°C
Liquid
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
KR-10
APC
KDELR1
KDELR1
A 21 residue synthetic peptide (amino acids 192-212) based on the bovine KDEL receptor and the peptide coupled to KLH.
100 μg
Primary
Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Hamster, Pig, Chicken, Chicken, Frog
Antibody
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt