missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ KDELR1 Antibody (SR2670), Novus Biologicals™
Rabbit Monoclonal Antibody
£259.00 - £307.00
Specifications
| Antigen | KDELR1 |
|---|---|
| Clone | SR2670 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041749
|
Novus Biologicals™
NBP3-27705-50ul |
50 μL |
£259.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041521
|
Novus Biologicals™
NBP3-27705-100ul |
100 μL |
£307.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KDELR1 Monoclonal antibody specifically detects KDELR1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| KDELR1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ER lumen protein retaining receptor 1, ERD2, ERD2.1KDEL receptor 1, HDEL, KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1, KDEL endoplasmic reticulum protein retention receptor 1, PM23, Putative MAPK-activating protein PM23 | |
| A synthesized peptide derived from human KDELR1 (Uniprot #: P24390) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR2670 | |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cellular Signaling, Chaperone Mediated Autophagy (CMA) | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 10945 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title