missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KDELC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566684
This item is not returnable.
View return policy
Description
Immunogen sequence: HENCDCPLQDS AAWLREMNCP ETIAQIQRDL AHFPAVDPEK IAVEIPKRFG QRQSLCHYTL KDNKVYIKTH GEHVG Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
Protein glucosyltransferase that catalyzes the transfer of glucose from UDP-glucose to a serine residue within the consensus sequence peptide C-X-N-T-X-G-S-F-X-C. Can also catalyze the transfer of xylose from UDP-xylose but less efficiently. Specifically targets extracellular EGF repeats of proteins such as NOTCH1 and NOTCH3. May regulate the transport of NOTCH1 and NOTCH3 to the plasma membrane and thereby the Notch signaling pathway.
Specifications
| KDELC1 | |
| Polyclonal | |
| Unconjugated | |
| KDELC1 | |
| endoplasmic reticulum resident protein 58; EP58; ER protein 58; ERp58; KDEL (Lys-Asp-Glu-Leu) containing 1; KDEL motif containing 1; KDEL motif-containing 1; KDEL motif-containing protein 1; KDEL1; KDELC1; POGLUT2; Protein O-glucosyltransferase 2; Protein O-xylosyltransferase POGLUT2; UNQ1910/PRO4357 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79070 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UW63 | |
| KDELC1 | |
| Recombinant Human KDELC1. Recombinant protein control fragment (Product #RP-107334). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction