missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDEL Antibody (1F2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61924H
This item is not returnable.
View return policy
Description
KDEL Monoclonal specifically detects KDEL in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| KDEL | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| KDELC1 | |
| KDEL peptide (C-AVKDEL -COOH) conjugated at the N-terminal to Bovine Serum Albumin (BSA) | |
| 100 μL | |
| Primary | |
| Detects KDEL tagged proteins. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| 1F2 | |
| Western Blot | |
| Endoplasmic reticulum resident protein 58, EP58KDEL1, ER protein 58, ERp58, KDEL (Lys-Asp-Glu-Leu) containing 1, KDEL motif-containing 1, KDEL motif-containing protein 1, MGC5302 | |
| Mouse | |
| Protein G purified | |
| Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
| 79070 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction