missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KD Scientific Stainless-Steel Syringes
Durable, electron-beam construction withstands high force, high pressure applications
Brand: KD Scientific 780817
Code : A0
Additional Details : Weight : 0.20000kg
Description
Eliminates hazards of glass syringe breakage.
- Compatible with most syringe pumps
- Fully autoclavable for reuse
- Adaptable to Luer Lock or Swagelok™ fittings
- Resistant to most chemicals
Specifications
50 mL | |
1-1/4 in. | |
28.4 mm | |
Stainless Steel (316) | |
1500 psi |
1-3/25 in. | |
31.7 mm | |
Syringe Pump | |
Luer-Lok Syringe |