Learn More
KD Scientific Stainless-Steel Syringes
Durable, electron-beam construction withstands high force, high pressure applications
Brand:  KD Scientific 780813
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15104184
  • £905.00 / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
Description
Description
Eliminates hazards of glass syringe breakage.
  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals
Specifications
Specifications
50 mL
1-1/4 in.
28.4 mm
Stainless Steel (316)
1500 psi
1-3/25 in.
31.7 mm
Syringe Pump
Swagelok 1/4 in. Syringe