missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCTD7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15873274
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15873274 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15873274 Supplier Invitrogen™ Supplier No. PA568478

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted to react with Human and Rat samples.

The KCTD gene family, including KCTD7, encode predicted proteins that contain N-terminal domain that is homologous to the T1 domain in voltage-gated potassium channels (see KCNA1; MIM 176260). KCTD7 displays a primary sequence and hydropathy profile indicating intracytoplasmic localization. EST database analysis showed that KCTD7 is expressed in human and mouse brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KCTD7
Gene Accession No. B1WC97, Q8BJK1, Q96MP8
Gene Alias 4932409E18; 9430010P06Rik; BTB/POZ domain-containing protein KCTD7; CLN14; EPM3; FLJ32069; KCTD7; potassium channel tetramerisation domain containing 7; potassium channel tetramerization domain containing 7
Gene Symbols KCTD7
Host Species Rabbit
Immunogen Synthesized peptide derived from human KCTD7 [aa 181-230].
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154881, 212919, 688993
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.