missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCTD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100587
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KCTD5.
KCTD5 is involved in interaction with CUL3 suggests that it may act as a substrate adapter in some E3 ligase complex. Does not affect the function of Kv channel Kv2.1/KCNB1, Kv1.2/KCNA2, Kv4.2/KCND2 and Kv3.4/KCNC4.
Specifications
| KCTD5 | |
| Polyclonal | |
| Unconjugated | |
| KCTD5 | |
| 2610030N08Rik; BTB/POZ domain-containing protein KCTD5; KCTD5; mKIAA0176; potassium channel tetramerisation domain containing 5; potassium channel tetramerization domain containing 5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 287109, 54442, 69259 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B5DEL1, Q8VC57, Q9NXV2 | |
| KCTD5 | |
| A synthesized peptide derived from human KCTD5(Accession Q9NXV2), corresponding to amino acid residues E7-T57. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction