missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KCTD5 Monoclonal antibody specifically detects KCTD5 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | KCTD5 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OTI3C8 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BTB/POZ domain-containing protein KCTD5, FLJ20040, potassium channel tetramerisation domain containing 5 |
| Host Species | Mouse |
| Immunogen | Full length recombinant protein of human KCTD5 (NP_061865) produced in HEK293T cells. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?