missing translation for 'onlineSavingsMsg'
Learn More

KCTD4 Antibody, Novus Biologicals™

Product Code. 18431752 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431752 0.1 mL 0.10mL
18462822 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18431752 Leverantör Novus Biologicals Leverantörsnummer NBP192045

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

KCTD4 Polyclonal antibody specifically detects KCTD4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen KCTD4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias bA321C24.3, BTB/POZ domain-containing protein KCTD4, potassium channel tetramerisation domain containing 4
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: LNFLRNGELLLPEGFRENQLLAQEAEFFQLKGLAEEVKSRWEKEQLTPRETTFLEITDNHDRSQGLRIFCNAPDF
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 386618
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.