missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44277
This item is not returnable.
View return policy
Description
Specifically detects KCTD3 in Human samples, and it is validated for Immunoprecipitation
KCTD3 Polyclonal specifically detects KCTD3 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| KCTD3 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| KCTD3 | |
| The epitope recognized by this antibody maps to a region between residue 675 to 725 of human BTB/POZ domain-containing protein KCTD3 using the numbering given in entry Q9Y597.2 (GeneID 51133). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
| Immunoprecipitation, Western Blot | |
| 1.0 mg/mL | |
| Immunoprecipitation 2 - 10 ug/mg lysate, Western Blot (Negative) | |
| BTB/POZ domain-containing protein KCTD3, potassium channel tetramerisation domain containing 3, renal carcinoma antigen NY-REN-45 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 51133 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction