missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCTD21 Recombinant Rabbit Monoclonal Antibody (K01_1T21)

Product Code. 30830422
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830422 50 μL
30826992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830422 Supplier Invitrogen™ Supplier No. MA564799
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

KCTD21 Recombinant Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD21
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1T21
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene KCTD21
Gene Accession No. Q3URF8, Q4G0X4
Gene Alias BTB/POZ domain-containing protein KCTD21; EG622320; K+ channel tetramerisation/BTB/POZ/BTB domain profile containing protein; KCASH2; KCASH2 protein; KCTD21; potassium channel tetramerisation domain containing 21; potassium channel tetramerization domain containing 21; Potassium channel tetramerization domain-containing protein 21
Gene Symbols KCTD21
Host Species Rabbit
Immunogen Recombinant protein of human KCTD21.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283219, 499209, 622320
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.