missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD19 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £375.00
Specifications
| Antigen | KCTD19 |
|---|---|
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18346116
|
Novus Biologicals
NBP3-04653-20UL |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18328734
|
Novus Biologicals
NBP3-04653-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCTD19 Polyclonal specifically detects KCTD19 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| KCTD19 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 146212 | |
| IgG | |
| Affinity purified |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| BTB/POZ domain-containing protein KCTD19, FLJ40162, potassium channel tetramerisation domain containing 19 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of mouse Kctd19 (NP_001288102.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title