missing translation for 'onlineSavingsMsg'
Learn More

KCTD15 Antibody (AT4C3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-49429

Product Code. 18227412

  • £358.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

KCTD15 Monoclonal specifically detects KCTD15 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KCTD15
Monoclonal
1 mg/mL
Western Blot 1:250-1:500, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:250
BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15
Mouse
Affinity Purified
RUO
79047
Human
IgG3 λ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunofluorescence
4C3
Unconjugated
NP_076981
KCTD15
Recombinant human KCTD15 (1-234aa) purified from E. coli.
0.1 mL
Primary
Human KCTD15 (1-234aa)
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only