missing translation for 'onlineSavingsMsg'
Learn More

KCTD15 Antibody (2B11), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18410277 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410277 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410277 Supplier Novus Biologicals Supplier No. NBP260232MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KCTD15 Monoclonal antibody specifically detects KCTD15 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD15
Applications Flow Cytometry
Classification Monoclonal
Clone 2B11
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15
Host Species Mouse
Immunogen Recombinant human KCTD15(1-159aa) purified from E. coli.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 79047
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.