missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD15 Antibody (2B11), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60232APC
This item is not returnable.
View return policy
Description
KCTD15 Monoclonal antibody specifically detects KCTD15 in Human samples. It is validated for Flow Cytometry
Specifications
| KCTD15 | |
| Monoclonal | |
| APC | |
| BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15 | |
| Recombinant human KCTD15(1-159aa) purified from E. coli. | |
| 0.1 mL | |
| metabolism | |
| 79047 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry | |
| 2B11 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction