missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD14 Antibody (OTI2E9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72171CL1
This item is not returnable.
View return policy
Description
KCTD14 Monoclonal antibody specifically detects KCTD14 in Human samples. It is validated for Western Blot
Specifications
| KCTD14 | |
| Monoclonal | |
| CoraFluor 1 | |
| BTB/POZ domain-containing protein KCTD14, MGC2376, potassium channel tetramerisation domain containing 14 | |
| Full length human recombinant protein of human KCTD14 (NP_076419) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| OTI2E9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 65987 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction