missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD14 Antibody (OTI1D11), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72173PECY7
This item is not returnable.
View return policy
Description
KCTD14 Monoclonal specifically detects KCTD14 in Human samples. It is validated for Flow Cytometry.
Specifications
| KCTD14 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| KCTD14 | |
| Full length recombinant protein of human KCTD14 (NP_076419) produced in HEK293T cells. | |
| 0.1 mL | |
| 65987.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI1D11 | |
| Flow Cytometry | |
| BTB/POZ domain-containing protein KCTD14, MGC2376, potassium channel tetramerisation domain containing 14 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction