missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15523-1-AP-150UL
This item is not returnable.
View return policy
Description
KCTD12 gene ontology annotations related to this gene include protein homooligomerization.Specifications
| KCTD12 | |
| Polyclonal | |
| Unconjugated | |
| KCTD12 | |
| C13orf2, KCTD12, KIAA1778, PFET1, PFETIN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 115207, 239217, 364458 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6WVG3, Q96CX2 | |
| Kctd12 | |
| KCTD12 Fusion Protein Ag5716 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction