missing translation for 'onlineSavingsMsg'
Learn More

KCTD12 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18416605 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416605 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416605 Supplier Novus Biologicals Supplier No. NBP180568AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KCTD12 Polyclonal antibody specifically detects KCTD12 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD12
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias BTB/POZ domain-containing protein KCTD12, C13orf2, chromosome 13 open reading frame 2, FLJ33073, KIAA1778PFET1PFETIN, Pfetin, potassium channel tetramerisation domain containing 12, Predominantly fetal expressed T1 domain
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 125 and 175 of human Predominantly Fetal Expressed T1 Domain using the numbering given in entry NP_612453.1 (GeneID 115207).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 115207
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.