missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD11 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72049
This item is not returnable.
View return policy
Description
KCTD11 Polyclonal specifically detects KCTD11 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| KCTD11 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| BTB/POZ domain-containing protein KCTD11, C17orf36, chromosome 17 open reading frame 36, MGC129844, potassium channel tetramerisation domain containing 11, REn, REN/KCTD11, RENpotassium channel tetramerization domain containing 11, retinoic acid, EGF, NGF induced gene protein, retinoic acid, EGF, NGF induced gene/potassium channel tetramerization domaincontaining 11 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 2 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_001002914.1 | |
| KCTD11 | |
| Peptide with sequence C-PEVEYGRLGLQP corresponding to internal region according to NP_001002914.1. | |
| 0.1 mg | |
| Signal Transduction | |
| 147040 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction