Learn More
Invitrogen™ KCTD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139857
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LGNTYILPKD SQVGPDVKSE AAPKRALYES VFGSGEICGP TSPKRLCIRP SEPVDAVVVV SVKHDPLPLL PEANGHRSTN SPT Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.
May repress the transcriptional activity of AP-2 family members, including TFAP2A, TFAP2B and TFAP2C to various extent.
Specifications
| KCTD1 | |
| Polyclonal | |
| Unconjugated | |
| KCTD1 | |
| BTB/POZ domain-containing protein KCTD1; C18orf5; KCTD1; potassium channel tetramerisation domain containing 1; potassium channel tetramerization domain containing 1; potassium channel tetramerization domain-containing protein 1; SENS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 284252 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q719H9 | |
| KCTD1 | |
| Recombinant protein corresponding to Human KCTD1. Recombinant protein control fragment (Product #RP-109022). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.