Learn More
Fisherbrand™ KCR Quick Coupling Kit
For fast connection and disconnection by simple snap-fastening. Fisherbrand™ KCR Quick Coupling Kits are designed for use with water faucets.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.17100kg
Description
- Comes with 15/21 tip and 20/37 adapter
- Facilitates the water filtration and purification process
Specifications
No | |
SI Wall Brackets | |
KCR | |
1 Kit |
For water purification | |
1-spigot, 1-fitting, 1 bi-conical quick coupling connection, 2 seals | |
4 bar | |
KCR Quick Coupling Kit |